accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14284-06-1

IUPAC Name: copper(2+) bis(1-ethoxy-1,3-dioxobutan-2-ide)
Synonyms: copper(2+) bis(1-ethoxy-1,3-dioxobutan-2-ide)
Molecular Weight (g/mol): 321.82
Molecular Formula: C12H18CuO6
InChi Key: OXFZSJOUPRCPJO-UHFFFAOYSA-N
SMILES: [Cu++].CCOC(=O)[CH-]C(C)=O.CCOC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)