accessibility menu, dialog, popup

UserName

Viewing profile as user:
ReReCCCCCCCCCCOOOOOOOOOO

CAS RN 14285-68-8

IUPAC Name: {[1,1,1,1,2,2,2,2,2-nonakis(hydroxymethyl)dirhenium-1-yl]methylidyne}-λ³-oxidane
Synonyms: {[1,1,1,1,2,2,2,2,2-nonakis(hydroxymethyl)dirhenium-1-yl]methylidyne}-λ³-oxidane
Molecular Weight (g/mol): 652.51
Molecular Formula: C10O10Re2
InChi Key: ZIZHEHXAMPQGEK-UHFFFAOYSA-N
SMILES: [O]#C[Re](C#[O])(C#[O])(C#[O])(C#[O])[Re](C#[O])(C#[O])(C#[O])(C#[O])C#[O]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)