accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142851-03-4

IUPAC Name: 1-tert-butyl 4-ethyl piperidine-1,4-dicarboxylate
Synonyms: 1-tert-butyl 4-ethyl piperidine-1,4-dicarboxylate
Molecular Weight (g/mol): 257.33
Molecular Formula: C13H23NO4
InChi Key: MYHJCTUTPIKNAT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CCN(CC1)C(=O)OC(C)(C)C
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)