accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1428551-28-3

NBr

CAS RN 1428551-28-3

IUPAC Name: 9-{[1,1'-biphenyl]-3-yl}-3-bromo-9H-carbazole
Synonyms: 9-{[1,1'-biphenyl]-3-yl}-3-bromocarbazole
Molecular Weight (g/mol): 398.3
Molecular Formula: C24H16BrN
InChi Key: NSRPRPVECXNOLB-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results