accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1429056-28-9

IUPAC Name: bis(2-oxa-7-azaspiro[3.5]nonane); oxalic acid
Synonyms: bis(2-oxa-7-azaspiro[3.5]nonane); oxalic acid
Molecular Weight (g/mol): 344.41
Molecular Formula: C16H28N2O6
InChi Key: WWVUFRRXXSVWBJ-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.C1OCC11CCNCC1.C1OCC11CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results