CAS RN 14294-10-1
IUPAC Name:
morpholine-4-carbothioamide
Synonyms:
morpholine-4-carbothioamide
Molecular Weight (g/mol):
146.21
Molecular Formula:
C5H10N2OS
InChi Key:
GSLBUBZXFUYMSW-UHFFFAOYSA-N
SMILES:
NC(=S)N1CCOCC1