accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SNHN

CAS RN 14294-11-2

IUPAC Name: (pyridin-2-yl)thiourea
Synonyms: pyridin-2-ylthiourea
Molecular Weight (g/mol): 153.2
Molecular Formula: C6H7N3S
InChi Key: SLUHLANJIVXTRQ-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)