accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNCH3

CAS RN 143-16-8

IUPAC Name: dihexylamine
Synonyms: 1-hexanamine, N-hexyl-
Molecular Weight (g/mol): 185.36
Molecular Formula: C12H27N
InChi Key: PXSXRABJBXYMFT-UHFFFAOYSA-N
SMILES: CCCCCCNCCCCCC
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)