accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOH

CAS RN 143-22-6

IUPAC Name: 2-[2-(2-butoxyethoxy)ethoxy]ethan-1-ol
Synonyms: triethylene glycol butyl ether
Molecular Weight (g/mol): 206.28
Molecular Formula: C10H22O4
InChi Key: COBPKKZHLDDMTB-UHFFFAOYSA-N
SMILES: CCCCOCCOCCOCCO
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)