CAS RN 14305-08-9
CAS RN 14305-08-9
IUPAC Name:
4,5-dibromo-2-phenyl-2,3-dihydropyridazin-3-one
Synonyms:
4,5-dibromo-2-phenylpyridazin-3-one
Molecular Weight (g/mol):
329.98
Molecular Formula:
C10H6Br2N2O
InChi Key:
NQJXEMRMTOSSBQ-UHFFFAOYSA-N
SMILES:
BrC1=C(Br)C(=O)N(N=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 4,5-Dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™
CAS: 14305-08-9 Molecular Formula: C10H6Br2N2O Molecular Weight (g/mol): 329.979 MDL Number: MFCD00052907 InChI Key: NQJXEMRMTOSSBQ-UHFFFAOYSA-N Synonym: 4,5-dibromo-2-phenylpyridazin-3 2h-one,4,5-dibromo-2-phenyl-2,3-dihydropyridazin-3-one,4,5-dibromo-2-phenyl-3 2h-pyridazinone,3 2h-pyridazinone, 4,5-dibromo-2-phenyl,4,5-dibromo-2-phenyl-3-pyridazinone,4,5-dibromo-2-phenyl-pyridazin-3-one,2-phenyl-4,5-dibromo-3 2h-pyridazinone,3 2h-pyridazinone,4,5-dibromo-2-phenyl,4,5-bis bromanyl-2-phenyl-pyridazin-3-one,4,5-dibromo-2-phenyl-2-hydropyridazin-3-one PubChem CID: 203396 IUPAC Name: 4,5-dibromo-2-phenylpyridazin-3-one SMILES: C1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Br)Br