accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHOHO

CAS RN 143060-31-5

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3,3-diphenylpropanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3,3-diphenylpropanoic acid
Molecular Weight (g/mol): 341.41
Molecular Formula: C20H23NO4
InChi Key: TYJDOLCFYZSNQC-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results