accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143071-39-0

IUPAC Name: 2-[(5-nitropyridin-2-yl)oxy]ethan-1-ol
Synonyms: 2-[(5-nitropyridin-2-yl)oxy]ethanol
Molecular Weight (g/mol): 184.15
Molecular Formula: C7H8N2O4
InChi Key: KESQFSZFUCZCEI-UHFFFAOYSA-N
SMILES: OCCOC1=NC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results