accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNONNOO

CAS RN 143112-51-0

IUPAC Name: 1-(7-nitro-2,1,3-benzoxadiazol-4-yl)pyrrolidin-3-amine
Synonyms: 1-(7-nitro-2,1,3-benzoxadiazol-4-yl)pyrrolidin-3-amine
Molecular Weight (g/mol): 249.23
Molecular Formula: C10H11N5O3
InChi Key: QSDDQXROWUJAJX-UHFFFAOYNA-N
SMILES: NC1CCN(C1)C1=CC=C(C2=NON=C12)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results