accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNI

CAS RN 143128-30-7

IUPAC Name: 1-[(4-iodophenyl)methyl]-1H-pyrazole
Synonyms: 1-[(4-iodophenyl)methyl]pyrazole
Molecular Weight (g/mol): 284.1
Molecular Formula: C10H9IN2
InChi Key: QRCIIHVBALOAQK-UHFFFAOYSA-N
SMILES: IC1=CC=C(CN2C=CC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results