accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14316-68-8

IUPAC Name: 2-methylbut-2-enoyl 2-methylbut-2-enoate
Synonyms: 2-methylbut-2-enoyl 2-methylbut-2-enoate
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: LIHLSLRANKCHLV-UHFFFAOYSA-N
SMILES: CC=C(C)C(=O)OC(=O)C(C)=CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results