accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOCH3

CAS RN 14318-66-2

IUPAC Name: 3-methoxy-N-methylaniline
Synonyms: 3-methoxy-N-methylaniline
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: ZFMZSZMUFWRAOG-UHFFFAOYSA-N
SMILES: CNC1=CC=CC(OC)=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results