CAS RN 14319-13-2
IUPAC Name:
tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) lanthanum
Synonyms:
tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) lanthanum
Molecular Weight (g/mol):
691.74
Molecular Formula:
C33H60LaO6
InChi Key:
VTNJXVDFRGBYJC-UHFFFAOYSA-N
SMILES:
[La].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C