accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143314-16-3

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 1-ethyl-3-methylimidazolium tetrafluoroborate
Molecular Weight (g/mol): 197.97
Molecular Formula: C6H11BF4N2
InChi Key: CUNYTRQQXKCRTJ-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCN1C=C[N+](C)=C1
Molecular Weight (g/mol) 
  • (6)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (7)
Percent Purity 
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (2)
  • (3)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (7)
  • (2)
  • (3)
  • (1)
  • (2)