accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143355-56-0

IUPAC Name: 3-hydroxy-7-methoxynaphthalene-2-carboxylic acid
Synonyms: 3-hydroxy-7-methoxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 218.21
Molecular Formula: C12H10O4
InChi Key: UDAQUPSJOGVPIX-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(O)C(=CC2=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results