accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OONCH3CH3

CAS RN 143356-43-8

IUPAC Name: N,N,N',N'-tetrapropylpropanediamide
Synonyms: N,N,N',N'-tetrapropylpropanediamide
Molecular Weight (g/mol): 270.42
Molecular Formula: C15H30N2O2
InChi Key: GXMHNVKZIWPXAP-UHFFFAOYSA-N
SMILES: CCCN(CCC)C(=O)CC(=O)N(CCC)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results