accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SCl

CAS RN 14345-97-2

IUPAC Name: 2-chloro-3-methylthiophene
Synonyms: 2-chloro-3-methylthiophene
Molecular Weight (g/mol): 132.61
Molecular Formula: C5H5ClS
InChi Key: KQFADYXPELMVHE-UHFFFAOYSA-N
SMILES: CC1=C(Cl)SC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results