accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNS

CAS RN 14346-25-9

IUPAC Name: 7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),2(6),9-trien-12-one
Synonyms: 7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),2(6),9-trien-12-one
Molecular Weight (g/mol): 192.24
Molecular Formula: C9H8N2OS
InChi Key: MZAZSAHKQBGEMA-UHFFFAOYSA-N
SMILES: O=C1NC=NC2=C1C1=C(CCC1)S2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results