accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClF

CAS RN 1435-44-5

IUPAC Name: 1-chloro-2,4-difluorobenzene
Synonyms: 1-chloro-2,4-difluorobenzene
Molecular Weight (g/mol): 148.54
Molecular Formula: C6H3ClF2
InChi Key: AJCSNHQKXUSMMY-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C(F)=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)