accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1435-46-7

FClCl

CAS RN 1435-46-7

IUPAC Name: 1,3-dichloro-5-fluorobenzene
Synonyms: 1,3-dichloro-5-fluorobenzene
Molecular Weight (g/mol): 164.99
Molecular Formula: C6H3Cl2F
InChi Key: BLWGKIXZAUOECS-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC(Cl)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results