accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClCl

CAS RN 1435-48-9

IUPAC Name: 2,4-dichloro-1-fluorobenzene
Synonyms: 2,4-dichloro-1-fluorobenzene
Molecular Weight (g/mol): 164.99
Molecular Formula: C6H3Cl2F
InChi Key: BDJZCCWUSOZUQG-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C=C1Cl
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)