accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrBr

CAS RN 1435-52-5

IUPAC Name: 1,4-dibromo-2-fluorobenzene
Synonyms: 1,4-dibromo-2-fluorobenzene
Molecular Weight (g/mol): 253.9
Molecular Formula: C6H3Br2F
InChi Key: WNSNPGHNIJOOPM-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC=C1Br
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results