accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14353-88-9

IUPAC Name: (2,3,4,5,6-pentafluorophenyl)[(2,2,2-trifluoroacetyl)oxy]-λ³-iodanyl 2,2,2-trifluoroacetate
Synonyms: (2,3,4,5,6-pentafluorophenyl)[(2,2,2-trifluoroacetyl)oxy]-λ³-iodanyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 519.99
Molecular Formula: C10F11IO4
InChi Key: OQWAXRPJEPTTSZ-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C([I](OC(=O)C(F)(F)F)OC(=O)C(F)(F)F)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results