accessibility menu, dialog, popup

UserName

Viewing profile as user:
AlOOOOOOFFFFFFFFFCH3CH3H3CHHH

CAS RN 14354-59-7

IUPAC Name: 2,8,13-trimethyl-4,10,15-tris(trifluoromethyl)-1λ³,5,7λ³,11,12λ³,16-hexaoxa-6-aluminaspiro[5.5⁶.5⁶]hexadeca-1,3,7,9,12,14-hexaen-6-yl
Synonyms: 2,8,13-trimethyl-4,10,15-tris(trifluoromethyl)-1λ³,5,7λ³,11,12λ³,16-hexaoxa-6-aluminaspiro[5.5⁶.5⁶]hexadeca-1,3,7,9,12,14-hexaen-6-yl
Molecular Weight (g/mol): 489.25
Molecular Formula: C15H15AlF9O6
InChi Key: DOYKOKXTRGISGV-DJFUMVPSSA-N
SMILES:
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results