accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1436-34-6

IUPAC Name: 2-butyloxirane
Synonyms: oxirane, butyl-
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: WHNBDXQTMPYBAT-UHFFFAOYNA-N
SMILES: CCCCC1CO1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)