accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 14360-50-0

IUPAC Name: 1-(furan-2-yl)hexan-1-one
Synonyms: 1-(furan-2-yl)hexan-1-one
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: YUAYWSBSIJVIBS-UHFFFAOYSA-N
SMILES: CCCCCC(=O)C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results