accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH2COOCH3

CAS RN 143633-91-4

IUPAC Name: 1,3-diethyl 2-(3-phenylprop-2-yn-1-yl)-2-(prop-2-en-1-yl)propanedioate
Synonyms: 1,3-diethyl 2-(3-phenylprop-2-yn-1-yl)-2-(prop-2-en-1-yl)propanedioate
Molecular Weight (g/mol): 314.38
Molecular Formula: C19H22O4
InChi Key: ZSPBIVGRCZUELN-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC=C)(CC#CC1=CC=CC=C1)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results