accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOONHNOCH3OH3CNH

CAS RN 143664-11-3

IUPAC Name: N-{4-[2-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)ethyl]phenyl}-5-methoxy-9-oxo-9,10-dihydroacridine-4-carboxamide
Synonyms: N-{4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]phenyl}-5-methoxy-9-oxo-10H-acridine-4-carboxamide
Molecular Weight (g/mol): 563.65
Molecular Formula: C34H33N3O5
InChi Key: OSFCMRGOZNQUSW-UHFFFAOYSA-N
SMILES: COC1=C2NC3=C(C=CC=C3C(=O)C2=CC=C1)C(=O)NC1=CC=C(CCN2CCC3=CC(OC)=C(OC)C=C3C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)