accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143688-82-8

IUPAC Name: 10-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)decanoic acid
Synonyms: 10-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}decanoic acid
Molecular Weight (g/mol): 409.53
Molecular Formula: C25H31NO4
InChi Key: VRQUPGNWTJGCBX-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCNC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)