accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN(E)

CAS RN 1437-15-6

IUPAC Name: 2-[(1E)-2-(pyridin-2-yl)ethenyl]pyridine
Synonyms: 1,2-(di-(A-pyridyl)ethylene
Molecular Weight (g/mol): 182.23
Molecular Formula: C12H10N2
InChi Key: HKEOCEQLCZEBMK-BQYQJAHWSA-N
SMILES: C(=C/C1=CC=CC=N1)\C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results