accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14379-00-1

IUPAC Name: disodium trioxocyclopent-1-ene-1,2-bis(olate)
Synonyms: disodium trioxocyclopent-1-ene-1,2-bis(olate)
Molecular Weight (g/mol): 186.03
Molecular Formula: C5Na2O5
InChi Key: OQXLFPHHAAAVKQ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results