accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1438-14-8

IUPAC Name: (2S)-2-(propan-2-yl)oxirane
Synonyms: (2S)-2-isopropyloxirane
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: REYZXWIIUPKFTI-RXMQYKEDSA-N
SMILES: CC(C)[C@H]1CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results