accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClOOHN

CAS RN 14389-06-1

IUPAC Name: 5-chloro-7-methyl-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 5-chloro-7-methyl-1H-indole-2,3-dione
Molecular Weight (g/mol): 195.6
Molecular Formula: C9H6ClNO2
InChi Key: LDFQLYHDZZPAGN-UHFFFAOYSA-N
SMILES: CC1=C2NC(=O)C(=O)C2=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results