accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1439-07-2

IUPAC Name: (2R,3R)-2,3-diphenyloxirane
Synonyms: R-trans-stilbene oxide (R,R)-stilbene oxide
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: ARCJQKUWGAZPFX-ZIAGYGMSSA-N
SMILES: O1[C@@H]([C@H]1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)