accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143983-91-9

IUPAC Name: indium(3+) trichloride
Synonyms: indium(III) chloride
Molecular Weight (g/mol): 221.17
Molecular Formula: Cl3In
InChi Key: PSCMQHVBLHHWTO-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[In+3]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)