accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHOHOOHONHHN

CAS RN 144-29-6

IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid; piperazine
Synonyms: citric acid; piperazine
Molecular Weight (g/mol): 278.26
Molecular Formula: C10H18N2O7
InChi Key: SWDXALWLRYIJHK-UHFFFAOYSA-N
SMILES: C1CNCCN1.OC(=O)CC(O)(CC(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results