accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaMgOONNOOOOOO

CAS RN 14402-88-1

IUPAC Name: magnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Synonyms: magnesium(2+) disodium ion(4-) edta
Molecular Weight (g/mol): 358.5
Molecular Formula: C10H12MgN2Na2O8
InChi Key: AWNVVAMWLMUZOZ-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Grade 
  • (1)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)