accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHN

CAS RN 144026-74-4

IUPAC Name: 5-(3-methoxyphenyl)-1H-pyrazole
Synonyms: 3-(3-methoxyphenyl)-2H-pyrazole
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: JAZQIOZMPNVSII-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results