CAS RN 14404-59-2
CAS RN 14404-59-2
IUPAC Name:
thulium(3+) trifluoride
Synonyms:
thulium(3+) trifluoride
Molecular Weight (g/mol):
225.93
Molecular Formula:
F3Tm
InChi Key:
AATUHDXSJTXIHB-UHFFFAOYSA-K
SMILES:
[F-].[F-].[F-].[Tm+3]
1
–
1
of
1
results
1
Thulium fluoride, 99.5% (REO), Thermo Scientific™
CAS: 14404-59-2 Molecular Formula: F3Tm Molecular Weight (g/mol): 225.929 InChI Key: AATUHDXSJTXIHB-UHFFFAOYSA-K Synonym: thulium fluoride,thulium 3+ ion trifluoride,acmc-20ajid,thulium iii ; fluoride PubChem CID: 10220162 IUPAC Name: thulium(3+);trifluoride SMILES: [F-].[F-].[F-].[Tm+3]