accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14405-43-7

IUPAC Name: gallium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: gallium acetylacetonate
Molecular Weight (g/mol): 367.05
Molecular Formula: C15H21GaO6
InChi Key: IMMRYVXLCAMRNK-UHFFFAOYSA-N
SMILES: [Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results