accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14405-49-3

IUPAC Name: iron(3+) tris(3-oxo-1,3-diphenylprop-1-en-1-olate)
Synonyms: iron(3+) tris(3-oxo-1,3-diphenylprop-1-en-1-olate)
Molecular Weight (g/mol): 725.6
Molecular Formula: C45H33FeO6
InChi Key: JWPHPBLJVLHKOZ-UHFFFAOYSA-K
SMILES: [Fe+3].[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results