CAS RN 1440509-01-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,4'-Bis(dodecyloxy)-3-methylazobenzene 93.0+%, TCI America™
CAS: 1440509-01-2 Molecular Formula: C37H60N2O2