accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1440509-03-4

IUPAC Name: [4-(hexyloxy)-3-methylphenyl][4-(hexyloxy)phenyl]diazene
Synonyms: [4-(hexyloxy)-3-methylphenyl][4-(hexyloxy)phenyl]diazene
Molecular Weight (g/mol): 396.58
Molecular Formula: C25H36N2O2
InChi Key: ULSNKXLZLMMAQJ-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(C=C1)N=NC1=CC(C)=C(OCCCCCC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results