accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 144052-40-4

IUPAC Name: {[3-(phosphonomethyl)phenyl]methyl}phosphonic acid
Synonyms: [3-(phosphonomethyl)phenyl]methylphosphonic acid
Molecular Weight (g/mol): 266.13
Molecular Formula: C8H12O6P2
InChi Key: PYWZCUXISXXGMI-UHFFFAOYSA-N
SMILES: OP(O)(=O)CC1=CC(CP(O)(O)=O)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results