accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 144053-98-5

IUPAC Name: (morpholin-2-yl)methanol hydrochloride
Synonyms: morpholin-2-ylmethanol hydrochloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: NBGXGDBTUJNTKJ-UHFFFAOYNA-N
SMILES: Cl.OCC1CNCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results