accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOOOHNHOO

CAS RN 144120-54-7

IUPAC Name: 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid
Synonyms: 4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid
Molecular Weight (g/mol): 409.44
Molecular Formula: C23H23NO6
InChi Key: ORKKMGRINLTBPC-UHFFFAOYNA-N
SMILES: OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)